Methyl-3-[2-(4-piperidinyl)ethoxy]benzoate hydrochloride

Supplier: Matrix Scientific

050048-500MG 050048-2.5G
101890-092EA 364.62 USD
101890-092 101890-094
Methyl-3-[2-(4-piperidinyl)ethoxy]benzoate hydrochloride
Methyl-3-[2-(4-piperidinyl)ethoxy]benzoate hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₂ClNO₃
MDL Number: MFCD13560450
CAS Number: 1220031-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR