3-Methoxyphenyl-2-(2-piperidinyl)ethyl ether hydrochloride

Supplier: Matrix Scientific

050046-500MG 050046-2.5G
101890-084EA 364.62 USD
101890-084 101890-086
3-Methoxyphenyl-2-(2-piperidinyl)ethyl ether hydrochloride
3-Methoxyphenyl-2-(2-piperidinyl)ethyl ether hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂ClNO₂
MDL Number: MFCD13560448
CAS Number: 1219949-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR