3-Methoxyphenyl-2-(4-piperidinyl)ethyl ether hydrochloride

Supplier: Matrix Scientific

050044-500MG 050044-2.5G
101890-076EA 364.62 USD
101890-076 101890-078
3-Methoxyphenyl-2-(4-piperidinyl)ethyl ether hydrochloride
3-Methoxyphenyl-2-(4-piperidinyl)ethyl ether hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂ClNO₂
MDL Number: MFCD13560446
CAS Number: 1220031-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR