N-[3-(3-Pyrrolidinylmethoxy)phenyl]acetamide hydrochloride

Supplier: Matrix Scientific

050043-500MG 050043-2.5G
101890-072EA 364.62 USD
101890-072 101890-074
N-[3-(3-Pyrrolidinylmethoxy)phenyl]acetamide hydrochloride
N-[3-(3-Pyrrolidinylmethoxy)phenyl]acetamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O₂
MDL Number: MFCD13560445
CAS Number: 1219949-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR