N-{3-[2-(2-Piperidinyl)ethoxy]phenyl}acetamide hydrochloride

Supplier: Matrix Scientific

050042-500MG 050042-2.5G
101890-068EA 364.62 USD
101890-068 101890-070
N-{3-[2-(2-Piperidinyl)ethoxy]phenyl}acetamide hydrochloride
N-{3-[2-(2-Piperidinyl)ethoxy]phenyl}acetamide hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃ClN₂O₂
MDL Number: MFCD13560444
CAS Number: 1219967-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR