N-{3-[2-(3-Piperidinyl)ethoxy]phenyl}acetamide hydrochloride

Supplier: Matrix Scientific

050041-500MG 050041-2.5G
101890-064EA 364.62 USD
101890-064 101890-066
N-{3-[2-(3-Piperidinyl)ethoxy]phenyl}acetamide hydrochloride
N-{3-[2-(3-Piperidinyl)ethoxy]phenyl}acetamide hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃ClN₂O₂
MDL Number: MFCD13560443
CAS Number: 1220037-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR