N-{3-[2-(4-Piperidinyl)ethoxy]phenyl}acetamide hydrochloride

Supplier: Matrix Scientific

050040-500MG 050040-2.5G
101890-060EA 364.62 USD
101890-060 101890-062
N-{3-[2-(4-Piperidinyl)ethoxy]phenyl}acetamide hydrochloride
N-{3-[2-(4-Piperidinyl)ethoxy]phenyl}acetamide hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃ClN₂O₂
MDL Number: MFCD13560442
CAS Number: 1219981-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR