3-[(3,4-Dichlorophenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050039-500MG 050039-2.5G
101890-056EA 364.62 USD
101890-056 101890-058
3-[(3,4-Dichlorophenoxy)methyl]pyrrolidine hydrochloride
3-[(3,4-Dichlorophenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄Cl₃NO
MDL Number: MFCD13560441
CAS Number: 1219980-73-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR