2-[2-(3,4-Dichlorophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050038-500MG 050038-2.5G
101890-052EA 364.62 USD
101890-052 101890-054
2-[2-(3,4-Dichlorophenoxy)ethyl]piperidine hydrochloride
2-[2-(3,4-Dichlorophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈Cl₃NO
MDL Number: MFCD13560440
CAS Number: 152009-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR