3-[2-(3,4-Dichlorophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050037-500MG 050037-2.5G
101890-048EA 364.62 USD
101890-048 101890-050
3-[2-(3,4-Dichlorophenoxy)ethyl]piperidine hydrochloride
3-[2-(3,4-Dichlorophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈Cl₃NO
MDL Number: MFCD13560439
CAS Number: 1220037-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR