4-[2-(3,4-Dichlorophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050036-500MG 050036-2.5G
101890-044EA 364.62 USD
101890-044 101890-046
4-[2-(3,4-Dichlorophenoxy)ethyl]piperidine hydrochloride
4-[2-(3,4-Dichlorophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈Cl₃NO
MDL Number: MFCD13560438
CAS Number: 158550-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR