3-[2-(3-Chlorophenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
050033-500MG
050033-2.5G
101890-032EA
364.62
USD
101890-032
101890-034
3-[2-(3-Chlorophenoxy)ethyl]piperidine hydrochloride
3-[2-(3-Chlorophenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₉Cl₂NO |
MDL Number:
MFCD13560435 CAS Number: 1219980-66-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...