4-[2-(3-Chlorophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050032-500MG 050032-2.5G
101890-028EA 364.62 USD
101890-028 101890-030
4-[2-(3-Chlorophenoxy)ethyl]piperidine hydrochloride
4-[2-(3-Chlorophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉Cl₂NO
MDL Number: MFCD13560434
CAS Number: 1219949-33-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR