3-[(3-Bromophenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050031-500MG 050031-2.5G
101890-024EA 364.62 USD
101890-024 101890-026
3-[(3-Bromophenoxy)methyl]pyrrolidine hydrochloride
3-[(3-Bromophenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅BrClNO
MDL Number: MFCD13560433
CAS Number: 1219982-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR