2-[2-(3-Bromophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050030-500MG 050030-2.5G
101890-020EA 364.62 USD
101890-020 101890-022
2-[2-(3-Bromophenoxy)ethyl]piperidine hydrochloride
2-[2-(3-Bromophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉BrClNO
MDL Number: MFCD13560432
CAS Number: 1219967-77-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR