3-[2-(3-Bromophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050029-500MG 050029-2.5G
101890-016EA 364.62 USD
101890-016 101890-018
3-[2-(3-Bromophenoxy)ethyl]piperidine hydrochloride
3-[2-(3-Bromophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉BrClNO
MDL Number: MFCD13560431
CAS Number: 1219976-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR