4-[2-(3-Bromophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050028-500MG 050028-2.5G
101890-012EA 364.62 USD
101890-012 101890-014
4-[2-(3-Bromophenoxy)ethyl]piperidine hydrochloride
4-[2-(3-Bromophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉BrClNO
MDL Number: MFCD13560430
CAS Number: 1220031-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR