3-[(2-Propylphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050027-500MG 050027-2.5G
101890-008EA 364.62 USD
101890-008 101890-010
3-[(2-Propylphenoxy)methyl]pyrrolidine hydrochloride
3-[(2-Propylphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂ClNO
MDL Number: MFCD13560429
CAS Number: 1219982-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR