2-[2-(2-Propylphenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
050026-500MG
050026-2.5G
101890-004EA
364.62
USD
101890-004
101890-006
2-[2-(2-Propylphenoxy)ethyl]piperidine hydrochloride
2-[2-(2-Propylphenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₆H₂₆ClNO |
MDL Number:
MFCD13560428 CAS Number: 1220029-15-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...