4-[2-(2-Propylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050024-500MG 050024-2.5G
101889-996EA 364.62 USD
101889-996 101889-998
4-[2-(2-Propylphenoxy)ethyl]piperidine hydrochloride
4-[2-(2-Propylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆ClNO
MDL Number: MFCD13560426
CAS Number: 1219949-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR