3-[(2-Allylphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050023-500MG 050023-2.5G
101889-992EA 364.62 USD
101889-992 101889-994
3-[(2-Allylphenoxy)methyl]pyrrolidine hydrochloride
3-[(2-Allylphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₀ClNO
MDL Number: MFCD13560425
CAS Number: 1220037-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR