2-[2-(2-Allylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050022-500MG 050022-2.5G
101889-988EA 364.62 USD
101889-988 101889-990
2-[2-(2-Allylphenoxy)ethyl]piperidine hydrochloride
2-[2-(2-Allylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₄ClNO
MDL Number: MFCD13560424
CAS Number: 1219960-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR