4-[2-(2-Allylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050020-500MG 050020-2.5G
101889-980EA 364.62 USD
101889-980 101889-982
4-[2-(2-Allylphenoxy)ethyl]piperidine hydrochloride
4-[2-(2-Allylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₄ClNO
MDL Number: MFCD13560422
CAS Number: 1219976-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR