3-[(2-Ethylphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050019-500MG 050019-2.5G
101889-976EA 364.62 USD
101889-976 101889-978
3-[(2-Ethylphenoxy)methyl]pyrrolidine hydrochloride
3-[(2-Ethylphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO
MDL Number: MFCD13560421
CAS Number: 1220031-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR