2-[2-(2-Ethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050018-500MG 050018-2.5G
101889-972EA 364.62 USD
101889-972 101889-974
2-[2-(2-Ethylphenoxy)ethyl]piperidine hydrochloride
2-[2-(2-Ethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD13560420
CAS Number: 1219949-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR