4-[2-(2-Ethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050016-500MG 050016-2.5G
101889-964EA 364.62 USD
101889-964 101889-966
4-[2-(2-Ethylphenoxy)ethyl]piperidine hydrochloride
4-[2-(2-Ethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD13560418
CAS Number: 1219976-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR