3-[(2,5-Dimethylphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050015-500MG 050015-2.5G
101889-960EA 364.62 USD
101889-960 101889-962
3-[(2,5-Dimethylphenoxy)methyl]pyrrolidine hydrochloride
3-[(2,5-Dimethylphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO
MDL Number: MFCD13560417
CAS Number: 1219982-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR