3-[2-(2,5-Dimethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050013-500MG 050013-2.5G
101889-952EA 364.62 USD
101889-952 101889-954
3-[2-(2,5-Dimethylphenoxy)ethyl]piperidine hydrochloride
3-[2-(2,5-Dimethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD13560415
CAS Number: 1219960-89-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR