4-[2-(2,5-Dimethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050012-500MG 050012-2.5G
101889-948EA 364.62 USD
101889-948 101889-950
4-[2-(2,5-Dimethylphenoxy)ethyl]piperidine hydrochloride
4-[2-(2,5-Dimethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD13560414
CAS Number: 1219976-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR