2-[2-(5-Isopropyl-2-methylphenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
050010-500MG
050010-2.5G
101889-940EA
364.62
USD
101889-940
101889-942
2-[2-(5-Isopropyl-2-methylphenoxy)ethyl]piperidine hydrochloride
2-[2-(5-Isopropyl-2-methylphenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₇H₂₈ClNO |
MDL Number:
MFCD13560412 CAS Number: 1220031-80-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...