3-[2-(5-Isopropyl-2-methylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050009-500MG 050009-2.5G
101889-936EA 364.62 USD
101889-936 101889-938
3-[2-(5-Isopropyl-2-methylphenoxy)ethyl]piperidine hydrochloride
3-[2-(5-Isopropyl-2-methylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₈ClNO
MDL Number: MFCD13560411
CAS Number: 1219960-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR