4-[2-(5-Isopropyl-2-methylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050008-500MG 050008-2.5G
101889-932EA 364.62 USD
101889-932 101889-934
4-[2-(5-Isopropyl-2-methylphenoxy)ethyl]piperidine hydrochloride
4-[2-(5-Isopropyl-2-methylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₈ClNO
MDL Number: MFCD13560410
CAS Number: 1219949-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR