3-[(2,4-Dimethylphenoxy)methyl]pyrrolidine hydrochloride
Supplier: Matrix Scientific
050007-500MG
050007-2.5G
101889-928EA
364.62
USD
101889-928
101889-930
3-[(2,4-Dimethylphenoxy)methyl]pyrrolidine hydrochloride
3-[(2,4-Dimethylphenoxy)methyl]pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₃H₂₀ClNO |
MDL Number:
MFCD13560409 CAS Number: 1219982-22-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...