2-[2-(2,4-Dimethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050006-500MG 050006-2.5G
101889-924EA 364.62 USD
101889-924 101889-926
2-[2-(2,4-Dimethylphenoxy)ethyl]piperidine hydrochloride
2-[2-(2,4-Dimethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD13560408
CAS Number: 1219976-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR