3-[2-(2,4-Dimethylphenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
050005-500MG
050005-2.5G
101889-920EA
364.62
USD
101889-920
101889-922
3-[2-(2,4-Dimethylphenoxy)ethyl]piperidine hydrochloride
3-[2-(2,4-Dimethylphenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₄ClNO |
MDL Number:
MFCD13560407 CAS Number: 1220019-97-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...