2-(4-Piperidinyl)ethyl-2,3,5-trimethylphenylether hydrochloride
Supplier: Matrix Scientific
050000-500MG
050000-2.5G
101889-900EA
363.08
USD
101889-900
101889-902
2-(4-Piperidinyl)ethyl-2,3,5-trimethylphenylether hydrochloride
2-(4-Piperidinyl)ethyl-2,3,5-trimethylphenylether hydrochloride
Hazard: Irritant
Formula:
C₁₆H₂₆ClNO |
MDL Number:
MFCD13560402 CAS Number: 1219949-27-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...