3-[2-(2,3-Dimethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049997-500MG 049997-2.5G
101889-888EA 364.62 USD
101889-888 101889-890
3-[2-(2,3-Dimethylphenoxy)ethyl]piperidine hydrochloride
3-[2-(2,3-Dimethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD13560399
CAS Number: 1220031-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR