4-[2-(2,3-Dimethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049996-500MG 049996-2.5G
101889-884EA 364.62 USD
101889-884 101889-886
4-[2-(2,3-Dimethylphenoxy)ethyl]piperidine hydrochloride
4-[2-(2,3-Dimethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD13560398
CAS Number: 1220019-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR