3-[(2-Methylphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

049995-500MG 049995-2.5G
101889-880EA 364.62 USD
101889-880 101889-882
3-[(2-Methylphenoxy)methyl]pyrrolidine hydrochloride
3-[(2-Methylphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈ClNO
MDL Number: MFCD13560397
CAS Number: 1219960-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR