2-[2-(2-Methylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049994-500MG 049994-2.5G
101889-876EA 364.62 USD
101889-876 101889-878
2-[2-(2-Methylphenoxy)ethyl]piperidine hydrochloride
2-[2-(2-Methylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂ClNO
MDL Number: MFCD13560396
CAS Number: 1219960-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR