3-{[2-(sec-Butyl)phenoxy]methyl}pyrrolidine hydrochloride
Supplier: Matrix Scientific
049991-500MG
049991-2.5G
101889-864EA
364.62
USD
101889-864
101889-866
3-{[2-(sec-Butyl)phenoxy]methyl}pyrrolidine hydrochloride
3-{[2-(sec-Butyl)phenoxy]methyl}pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₄ClNO |
MDL Number:
MFCD13560393 CAS Number: 1219972-76-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...