2-{2-[2-(sec-Butyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049990-500MG 049990-2.5G
101889-860EA 364.62 USD
101889-860 101889-862
2-{2-[2-(sec-Butyl)phenoxy]ethyl}piperidine hydrochloride
2-{2-[2-(sec-Butyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₈ClNO
MDL Number: MFCD13560392
CAS Number: 1220031-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR