3-{2-[2-(sec-Butyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049989-500MG 049989-2.5G
101889-856EA 364.62 USD
101889-856 101889-858
3-{2-[2-(sec-Butyl)phenoxy]ethyl}piperidine hydrochloride
3-{2-[2-(sec-Butyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₈ClNO
MDL Number: MFCD13560391
CAS Number: 1219949-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR