3-[(2-Isopropylphenoxy)methyl]pyrrolidine hydrochloride
Supplier: Matrix Scientific
049987-500MG
049987-2.5G
101889-848EA
364.62
USD
101889-848
101889-850
3-[(2-Isopropylphenoxy)methyl]pyrrolidine hydrochloride
3-[(2-Isopropylphenoxy)methyl]pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₂ClNO |
MDL Number:
MFCD13560389 CAS Number: 1220028-81-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...