2-[2-(2-Isopropylphenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
049986-500MG
049986-2.5G
101889-844EA
364.62
USD
101889-844
101889-846
2-[2-(2-Isopropylphenoxy)ethyl]piperidine hydrochloride
2-[2-(2-Isopropylphenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₆H₂₆ClNO |
MDL Number:
MFCD13560388 CAS Number: 1219949-24-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...