3-[2-(2-Isopropylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049985-500MG 049985-2.5G
101889-840EA 364.62 USD
101889-840 101889-842
3-[2-(2-Isopropylphenoxy)ethyl]piperidine hydrochloride
3-[2-(2-Isopropylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆ClNO
MDL Number: MFCD13560387
CAS Number: 1220019-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR