4-[2-(2-Isopropylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049984-500MG 049984-2.5G
101889-836EA 364.62 USD
101889-836 101889-838
4-[2-(2-Isopropylphenoxy)ethyl]piperidine hydrochloride
4-[2-(2-Isopropylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆ClNO
MDL Number: MFCD13560386
CAS Number: 1220028-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR