2-(tert-Butyl)phenyl-3-pyrrolidinylmethyl ether hydrochloride

Supplier: Matrix Scientific

049983-500MG 049983-2.5G
101889-832EA 364.62 USD
101889-832 101889-834
2-(tert-Butyl)phenyl-3-pyrrolidinylmethyl ether hydrochloride
2-(tert-Butyl)phenyl-3-pyrrolidinylmethyl ether hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD13560385
CAS Number: 1146960-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR