4-{2-[2-(tert-Butyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049980-500MG 049980-2.5G
101889-820EA 364.62 USD
101889-820 101889-822
4-{2-[2-(tert-Butyl)phenoxy]ethyl}piperidine hydrochloride
4-{2-[2-(tert-Butyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₈ClNO
MDL Number: MFCD13560382
CAS Number: 1146960-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR