3-{[2-(Trifluoromethyl)phenoxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

049979-500MG 049979-2.5G
101889-816EA 364.62 USD
101889-816 101889-818
3-{[2-(Trifluoromethyl)phenoxy]methyl}pyrrolidine hydrochloride
3-{[2-(Trifluoromethyl)phenoxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₅ClF₃NO
MDL Number: MFCD13560381
CAS Number: 1220028-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR